missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ mCherry Monoclonal Antibody (16D7), Alexa Fluor™ 647
Description
mCherry Monoclonal Antibody for Flow, ICC/IF, Western Blot
MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.
Specifications
Specifications
| Antigen | mCherry |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 16D7 |
| Conjugate | Alexa Fluor 647 |
| Formulation | PBS with 5mM sodium azide; pH 7.5 |
| Gene Alias | mCherry |
| Host Species | Rat |
| Immunogen | Full-length mCherry was used as the immunogen. |
| Purification Method | Affinity chromatography |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?