missing translation for 'onlineSavingsMsg'
Learn More

mCherry Monoclonal Antibody (16D7), Alexa Fluor™ 488, Invitrogen™

Rat Monoclonal Antibody

Brand:  Invitrogen M11239

Code : NEW

Additional Details : Weight : 0.25000kg

Product Code. 15421194

  • £256.00 / 1 set
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.
Specifications

Specifications

mCherry
Monoclonal
Alexa Fluor 488
mCherry
Full-length mCherry was used as the immunogen
50 Tests
Primary
Antibody
IgG2a
Flow Cytometry
16D7
PBS with 5mM sodium azide; pH 7.5
Rat
Affinity chromatography
RUO
4° C
Liquid
Tag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers