Learn More
mCherry Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-43720
This item is not returnable.
View return policy
Description
Specifically detects mCherry in Human, Other, Zebrafish samples, and it is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
mCherry Polyclonal specifically detects mCherry in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount.
Specifications
| Unconjugated | |
| DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein | |
| Immunogen affinity purified |
| Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Frozen, Proximity Ligation Assay, Immunohistochemistry 1:100-1:500 | |
| 27 kDa |