missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mCherry Antibody, Novus Biologicals™
Description
mCherry Polyclonal antibody specifically detects mCherry in Nonspecies specific samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | mCherry |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000, Immunocytochemistry/ Immunofluorescence 1:1000 |
| Formulation | 50% PBS, 50% glycerol |
| Gene Alias | DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein |
| Host Species | Goat |
| Immunogen | This mCherry Antibody was developed against full length recombinant mCherry protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?