missing translation for 'onlineSavingsMsg'
Learn More

mCherry Antibody, Novus Biologicals™

Product Code. 18691428
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18691428 50 μg 50µL
18697597 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18691428 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP30555850ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

mCherry Polyclonal antibody specifically detects mCherry in Nonspecies specific samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen mCherry
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunocytochemistry/ Immunofluorescence 1:1000
Formulation 50% PBS, 50% glycerol
Gene Alias DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein
Host Species Goat
Immunogen This mCherry Antibody was developed against full length recombinant mCherry protein
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Fusion Proteins
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.