missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mCherry Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£236.00 - £468.00
Specifications
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
|---|---|
| Isotype | IgG |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18709643
|
Novus Biologicals
NBP2-25157SS |
£236.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18025544
|
Novus Biologicals
NBP2-25157 |
0.1 mL |
£468.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
mCherry Polyclonal specifically detects mCherry in Non-species specific samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Live Imaging Microscopy, Knockdown Validated, Fluorescence Imaging.Specifications
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| IgG | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title