missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mCherry Antibody, Novus Biologicals™
Chicken Polyclonal Antibody has been used in 26 publications
Brand: Novus Biologicals NBP2-25158-0.025ML
This item is not returnable.
View return policy
Description
mCherry Polyclonal specifically detects mCherry in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount.
Specifications
| mCherry | |
| Polyclonal | |
| Western Blot 1:2000-1:5000, ELISA, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, IHC | |
| DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein | |
| Full length recombinant protein expressed in and purified from E. coli. | |
| Ammonium sulfate precipitation | |
| RUO | |
| This antibody detects mCherry. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgY |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Free Floating), Immunofluorescence, Western Blot | |
| Unconjugated | |
| Antibody is supplied as an aliquot of concentrated IgY preparation. with 5mM Sodium Azide | |
| Chicken | |
| 28 kDa | |
| 0.025 mL | |
| Primary | |
| All species | |
| Precipitation |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction