missing translation for 'onlineSavingsMsg'
Learn More

mCherry Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18419856 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419856 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419856 Supplier Novus Biologicals Supplier No. NBP225157MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

mCherry Polyclonal antibody specifically detects mCherry in Nonspecies specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Live Imaging Microscopy, Immunohistochemistry, KnockDown, Fluorescence Imaging
TRUSTED_SUSTAINABILITY

Specifications

Antigen mCherry
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Live Imaging Microscopy, Immunohistochemistry (Whole-Mount), KnockDown, Fluorescence Imaging
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Live Imaging Microscopy, Immunohistochemistry Whole-Mount, Knockdown Validated, Fluorescence Imaging
Formulation 50mM Sodium Borate
Gene Alias DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein
Host Species Rabbit
Immunogen This mCherry Antibody was developed against full length recombinant mCherry protein
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Fusion Proteins
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.