missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mCherry Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-25157AF532
This item is not returnable.
View return policy
Description
mCherry Polyclonal specifically detects mCherry in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| mCherry | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated | |
| DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein | |
| This mCherry Antibody was developed against full length recombinant mCherry protein | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Rabbit | |
| 27 kDa | |
| 0.1 mL | |
| Cellular Markers, Fusion Proteins | |
| Non-species specific, Mouse, Rat, Bacteria | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction