missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mCherry Antibody (2783C), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB11041F
This item is not returnable.
View return policy
Description
mCherry Monoclonal antibody specifically detects mCherry in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunocytochemistry
Specifications
| mCherry | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| 2783C | |
| Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein | |
| Human mCherry synthetic peptide | |
| 0.1 mL | |
| Cellular Markers, Fusion Proteins | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction