missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCG10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-76729
This item is not returnable.
View return policy
Description
MCG10 Polyclonal specifically detects MCG10 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| MCG10 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| alpha-CP4, alphaCP-4 protein, CBP, LIP4, LYST-interacting protein, MCG10, poly(rC) binding protein 4, poly(rC)-binding protein 4, RNA binding protein MCG10 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1 mg/ml | |
| Western Blot 0.5 - 2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1-20 μg/mL, Immunoprecipitation | |
| EAW65166 | |
| PCBP4 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human MCG10. The immunogen is located within the last 50 amino acids of MCG10. Amino Acid Squence: AAANGSKKAERQKFSPY | |
| 0.1 mg | |
| Apoptosis, Autophagy, Cancer, DNA Repair, Embryonic Stem Cell Markers, Stem Cell Markers | |
| 57060 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction