missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCFD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13068-1-AP-150UL
This item is not returnable.
View return policy
Description
Multiple coagulation factor deficiency protein 2 (MCFD2) is localized in the endoplasmic reticulum-Golgi intermediate compartment (ERGIC) through a direct, calcium-dependent interaction with LMAN1. The MCFD2-LMAN1 complex forms a specific cargo receptor for the transport of selected proteins from the endoplasmic reticulum to the Golgi apparatus. Mutations in the MCFD2 gene may cause of factor V and factor VIII combined deficiency (F5F8D). F5F8D is an autosomal recessive human bleeding disorder characterized by the reduction of both clotting proteins.Specifications
| MCFD2 | |
| Polyclonal | |
| Unconjugated | |
| MCFD2 | |
| LMAN1IP, MCFD2, SDNSF | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 193813, 246117, 90411 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8K5B2, Q8K5B3, Q8NI22 | |
| MCFD2 | |
| MCFD2 Fusion Protein Ag3768 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction