missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCFD2 Monoclonal Antibody (294301)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524000
This item is not returnable.
View return policy
Description
In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse SDNSF is observed. In Western blots, no cross-reactivity with recombinant mouse SDNSF is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Multiple coagulation factor deficiency protein 2 (MCFD2) is localized in the endoplasmic reticulum-Golgi intermediate compartment (ERGIC) through a direct, calcium-dependent interaction with LMAN1. The MCFD2-LMAN1 complex forms a specific cargo receptor for the transport of selected proteins from the endoplasmic reticulum to the Golgi apparatus. Mutations in the MCFD2 gene may cause of factor V and factor VIII combined deficiency (F5F8D). F5F8D is an autosomal recessive human bleeding disorder characterized by the reduction of both clotting proteins.
Specifications
| MCFD2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8NI22 | |
| Mcfd2 | |
| E. coli-derived recombinant human SDNSF/MCFD2 Glu27-Gln146. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry, Western Blot | |
| 294301 | |
| Unconjugated | |
| Mcfd2 | |
| 1810021C21Rik; F5F8D; F5F8D2; LMAN1IP; MCFD2; multiple coagulation factor deficiency 2; multiple coagulation factor deficiency protein 2; Multiple coagulation factor deficiency protein 2 homolog; neural stem cell derived neuronal survival protein; neural stem cell-derived neuronal survival protein; Sdnsf; stem cell derived neuronal survival protein | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 90411 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction