missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCF2L Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522223
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (90%), Rat (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a guanine nucleotide exchange factor that interacts specifically with the GTP-bound Rac1 and plays a role in the Rho/Rac signaling pathways. A variant in this gene was associated with osteoarthritis. Alternative splicing results in multiple transcript variants.
Specifications
| MCF2L | |
| Polyclonal | |
| Unconjugated | |
| MCF2L | |
| ARHGEF14; DBL's big sister; DBS; FLJ12122; Guanine nucleotide exchange factor DBS; KIAA0362; mcf 2l; MCF.2 cell line derived transforming sequence like; MCF.2 cell line derived transforming sequence-like; mcf.2 transforming sequence-like; MCF2 transforming sequence-like protein; MCF2L; MCF2-transforming sequence-like protein; OST; OST oncogene; RP11-98F14.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23263 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O15068 | |
| MCF2L | |
| Recombinant protein encompassing a sequence within the N-terminus region of human Dbs. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction