missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCF2 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00004168-B01P-50ug
This item is not returnable.
View return policy
Description
MCF2 Polyclonal antibody specifically detects MCF2 in Human samples. It is validated for Western Blot
Specifications
| MCF2 | |
| Polyclonal | |
| Western Blot | |
| AAI60052.1 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| ARHGEF21, DBL, MCF 2, MCF.2 cell line derived transforming sequence, MCF-2, MCF2 MCF.2 cell line derived transforming sequence | |
| MCF2 (AAI60052.1, 1 a.a. - 985 a.a.) full-length human protein. MQDIAFLSGGRGKDNAWIITFPENCNFRCIPEEVIAKVLTYLTSIARQNGSDSRFTIILDRRLDTWSSLKISLQKISASFPGNLHLVLVLRPTSFLQRTFTDIGFWFSQEDFMLKLPVVMLSSVSDLLTYIDDKQLTPELGGTLQYCHSEWIIFRNAIENFALTVKEMAQMLQSFGTELAETELPDDIPSIEEILAIRAERYHLLKNDITAVTKEGKILLTNLEVPDTEGAVSSRLECHRQISGDWQTINKLLTQVHDMETAFDGFWEKHQLKMEQYLQLWKFEQDFQQLVTEVEFLLNQQAELADVTGTIAQVKQKIKKLENLDENSQELLSKAQFVILHGHKLAANHHYALDLICQRCNELRYLSDILVNEIKAKRIQLSRTFKMHKLLQQARQCCDEGECLLANQEIDKFQSKEDAQKALQDIENFLEMALPFINYEPETLQYEFDVILSPELKVQMKTIQLKLENIRSIFENQQAGFRNLADKHVRPIQFVVPTPENLVTSGTPFFSSKQGKKTWRQNQSNLKIEVVPDCQEKRSSGPSSSLDNGNSLDVLKNHVLNELIQTERVYVRELYTVLLGYRAEMDNPEMFDLMPPLLRNKKDILFGNMAEIYEFHNDIFLSSLENCAHAPERVGPCFLERKDDFQMYAKYCQNKPRSETIWRKYSECAFFQECQRKLKHRLRLDSYLLKPVQRITKYQLLLKELLKYSKDCEGSALLKKALDAMLDLLKSVNDSMHQIAINGYIGNLNELGKMIMQGGFSVWIGHKKGATKMKDLARFKPMQRHLFLYEKAIVFCKRRVESGEGSDRYPSYSFKHCWKMDEVGITEYVKGDNRKFEIWYGEKEEVYIVQASNVDVKMTWLKEIRNILLKQQELLTVKKRKQQDQLTERDKFQISLQQNDEKQQGAFISTEETELEHTSTVVEVCEAIASVQAEANTVWTEASQSAEISEEPAEWSSNYFYPTYDENEEENRPLMRPVSEMALLY | |
| 50 μg | |
| Cancer, GPCR, Signal Transduction | |
| 4168 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction