missing translation for 'onlineSavingsMsg'
Learn More

MCEMP1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA553223

Additional Details : Weight : 0.02000kg

Product Code. 15701171

  • £441.00 / 100µL
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: MSKELLGFKR ELWNVSNSVQ ACEERQKRGW DSVQQSITMV RSKIDRLETT LAGIKNIDTK VQKILEVLQK MPQSSPQ Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 37%.

MCEMP1 Polyclonal antibody specifically detects MCEMP1 in Human samples. It is validated for Immunohistochemistry, Western Blot.
Specifications

Specifications

MCEMP1
Polyclonal
Unconjugated
MCEMP1
C19orf59; mast cell expressed membrane protein 1; mast cell-expressed membrane protein 1; MCEMP1; MGC132456
Rabbit
Antigen affinity chromatography
RUO
199675
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8IX19
MCEMP1
Recombinant protein corresponding to Human MCEMP1
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only