missing translation for 'onlineSavingsMsg'
Learn More

MCEMP1 Antibody [DyLight 594], Novus Biologicals™

Product Code. 30738438 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30738438 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738438 Supplier Novus Biologicals Supplier No. AF4994M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

MCEMP1 Polyclonal antibody specifically detects MCEMP1 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCEMP1
Applications Flow Cytometry, Western Blot, CyTOF
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias C19orf59, chromosome 19 open reading frame 59, mast cell-expressed membrane protein 1, MCEMP1, MGC132456
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human MCEMP1, Val106-Gln187, Accession # Q8IX19
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199675
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.