missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCEE Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-04474-20UL
This item is not returnable.
View return policy
Description
MCEE Polyclonal specifically detects MCEE in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MCEE | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84693 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| DL-methylmalonyl-CoA racemase, EC 5.1.99.1, GLOD2, glyoxalase domain containing 2, methylmalonyl CoA epimerase, methylmalonyl-CoA epimerase, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-176 of human MCEE (NP_115990.3). | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction