missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MCCC1 Polyclonal specifically detects MCCC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | MCCC1 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | 3-methylcrotonyl-CoA carboxylase 1, DKFZp686B20267, EC 6.4.1, EC 6.4.1.4, MCCAFLJ25545, MCC-B, methylcrotonoyl-CoA carboxylase 1 (alpha), methylcrotonoyl-Coenzyme A carboxylase 1 (alpha), mitochondrial |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 526-725 of human MCCC1 (NP_064551.3). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?