missing translation for 'onlineSavingsMsg'
Learn More

MCC950 sodium, MedChemExpress

Product Code. 16485368
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485368 10 mg 10mg
16468087 5 mg 5mg
16445996 100 mg 100mg
16421567 50 mg 50mg
16421056 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16485368 Supplier Medchem Express Supplier No. HY12815A
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress MCC950 sodium (CP-456773 sodium; CRID3 sodium salt) is a potent, selective NLRP3 inhibitor with IC50s of 7.5 and 8.1 nM in BMDMs and HMDMs, respectively.

MCC950 sodium (CP-456773 sodium

Specifications

Chemical Name or Material MCC950 sodium
CAS 256373-96-3
Color White
Purity Grade Notes Research
Molecular Formula C20H23N2NaO5S
Quantity 10 mg
Synonym CP-456773 sodium CRID3 sodium salt
Solubility Information DMSO : 100 mg/mL (234.49 mM; Need ultrasonic) ∣H2O : ≥ 30 mg/mL (70.35 mM)
SMILES O=S(C1=CC(C(C)(O)C)=CO1)([N-]C(NC2=C3CCCC3=CC4=C2CCC4)=O)=O.[Na+]
Molecular Weight (g/mol) 426.46
Formula Weight 426.46
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.