missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11525-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a candidate colorectal tumor suppressor gene that is thought to negatively regulate cell cycle progression. The orthologous gene in the mouse expresses a phosphoprotein associated with the plasma membrane and membrane organelles, and overexpression of the mouse protein inhibits entry into S phase. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| MCC | |
| Polyclonal | |
| Unconjugated | |
| MCC | |
| MCC, MCC1, mutated in colorectal cancers, Protein MCC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 328949, 4163 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P23508 | |
| Mcc | |
| MCC Fusion Protein Ag2089 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction