missing translation for 'onlineSavingsMsg'
Learn More

MCC Antibody, R&D Systems™

Product Code. 18604428 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18604428 0.1 mL 0.10mL
18661979 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18604428 Supplier Novus Biologicals Supplier No. NBP248671
  • ONLINE EXCLUSIVE PRICE
    £490.45
    £418.95 / 0.1mL
    Save £71.50
    14% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

MCC Polyclonal antibody specifically detects MCC in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCC
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias colorectal mutant cancer protein, DKFZp762O1615, FLJ38893, FLJ46755, MCC1, mutated in colorectal cancers, Protein MCC
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: VQELVSALERLTKSSEIRHQQSAEFVNDLKRANSNLVAAYEKAKKKHQNKLKKLESQMMAMVERHETQVRMLKQRIALLEEENSRPHTNETS
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4163
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.