missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113115
This item is not returnable.
View return policy
Description
MCAT Polyclonal Antibody for Western Blot, IHC (P), ELISA
Catalyzes the transfer of a malonyl moiety from malonyl-CoA to the free thiol group of the phosphopantetheine arm of the mitochondrial ACP protein (NDUFAB1). This suggests the existence of the biosynthesis of fatty acids in mitochondrias.
Specifications
| MCAT | |
| Polyclonal | |
| Unconjugated | |
| MCAT | |
| [Acyl-carrier-protein] malonyltransferase; [acyl-carrier-protein] S-malonyltransferase; AI225907; BC025519; fabD; FASN2C; malonyl CoA:ACP acyltransferase (mitochondrial); malonyl CoA-acyl carrier protein transacylase, mitochondrial; malonyl-CoA:acyl carrier protein transacylase, mitochondrial; malonyl-CoA-acyl carrier protein transacylase; malonyl-CoA-acyl carrier protein transacylase, mitochondrial; Mcat; MCT; mitochondrial malonyl CoA:ACP acyltransferase; Mitochondrial malonyltransferase; Mt; NET62; RGD1306698 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 223722, 27349, 315173 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8IVS2, Q8R3F5 | |
| MCAT | |
| Recombinant Human Malonyl-CoA-acyl carrier protein transacylase, mitochondrial protein (22-117AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction