missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCAT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35607-100ul
This item is not returnable.
View return policy
Description
MCAT Polyclonal antibody specifically detects MCAT in Human samples. It is validated for ELISA,Western Blot
Specifications
| MCAT | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 2.3.1.39, fabD, FASN2C, malonyl CoA:ACP acyltransferase (mitochondrial), malonyl-CoA:acyl carrier protein transacylase, mitochondrial, malonyl-CoA-acyl carrier protein transacylase, mitochondrial, MCT[Acyl-carrier-protein] malonyltransferase, Mitochondrial malonyltransferase, MTMGC47838, NET62 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 191-390 of human MCAT (NP_775738.3).,, Sequence:, SGMLSVLGQPQSKFNFACLEAREHCKSLGIENPVCEVSNYLFPDCRVISGHQEALRFLQKNSSKFHFRRTRMLPVSGAFHTRLMEPAVEPLTQALKAVDIKKPLVSVYSNVHAHRYRHPGHIHKLLAQQLVSPVKWEQTMHAIYERKKGRGFPQTFEVGPGRQLGAILKSCNMQAWKSYSAVDVLQTLEHVDLDPQEPPR | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 27349 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction