Learn More
MCAM/CD146 Antibody (CL12855) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18546JF646
Product Code. 30297939
  • £582.00 / 0.10mL
Description
MCAM/CD146 Monoclonal antibody specifically detects MCAM/CD146 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MCAM/CD146
Monoclonal
Janelia Fluor 646
CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin
This antibody was developed using a recombinant protein derived from P43121, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
4162
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL12855
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified