missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCAM/CD146 Antibody (C146/634) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
MCAM/CD146 Monoclonal antibody specifically detects MCAM/CD146 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Specifications
Specifications
| Antigen | MCAM/CD146 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
| Clone | C146/634 |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml |
| Formulation | 10 mM PBS |
| Gene Alias | CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?