missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCAM/CD146 Antibody (733216), PE/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB7718PECY7
This item is not returnable.
View return policy
Description
MCAM/CD146 Monoclonal antibody specifically detects MCAM/CD146 in Mouse samples. It is validated for Flow Cytometry
Specifications
| MCAM/CD146 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 733216 | |
| Flow Cytometry | |
| CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin | |
| Mouse myeloma cell line NS0-derived recombinant mouse MCAM/CD146, Met1-Val563, Accession # Q8R2Y2 | |
| 0.1 mL | |
| Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
| 4162 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction