missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCAM/CD146 Antibody (012), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89202V
This item is not returnable.
View return policy
Description
MCAM/CD146 Monoclonal specifically detects MCAM/CD146 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MCAM/CD146 | |
| Monoclonal | |
| DyLight 405 | |
| CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human MCAM/CD146 extracellular domain (NP_006491.2; Met 1-Gly 559) . | |
| 0.1 mL | |
| Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
| 4162 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 12 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human CD66a Human CD166 Human CD106 Human BCAM Human CD226 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction