missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCAK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527833
This item is not returnable.
View return policy
Description
Recommended positive controls: K562. Predicted reactivity: Mouse (80%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC), which consists of 60-100 proteins. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion while larger molecules are transported by an active process. The protein encoded by this gene belongs to the importin alpha family, and is involved in nuclear protein import.
Specifications
| MCAK | |
| Polyclonal | |
| Unconjugated | |
| KIF2C | |
| 4930402F02Rik; CT139; ESTM5; hypothetical protein; KIF2C; kinesin family member 2C; kinesin-like 6; kinesin-like protein 6; Kinesin-like protein KIF2C; kinesin-related protein 2; KNSL6; KRP2; MCAK; Mitotic centromere-associated kinesin; QtsA-16015; RP11-269F19.1; testis tissue sperm-binding protein Li 68n; X83316 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11004 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q99661 | |
| KIF2C | |
| Recombinant protein encompassing a sequence within the center region of human MCAK. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction