missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCAF1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | MCAF1 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MCAF1 Polyclonal specifically detects MCAF1 in Mouse samples. It is validated for Western Blot.Specifications
| MCAF1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| activating transcription factor 7 interacting protein, activating transcription factor 7-interacting protein 1, ATF7-interacting protein, ATFa-associated modulator, ATF-interacting protein, ATF-IP, FLJ10139, FLJ10688, hAM, MBD1-containing chromatin associated factor, MCAF1AM, MCAFMBD1-containing chromatin-associated factor 1, MMCP 6, MMCP6, MMCP-6, P621 | |
| The immunogen is a synthetic peptide directed towards the middle region of Mouse MCAF1 (NP_062299). Peptide sequence MSTPEGEKSEKDGKAEEEERVPAEEQPPVRNEFSRRKRSKSEDMDSVESK | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 55729 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title