missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCAF1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97607G
This item is not returnable.
View return policy
Description
MCAF1 Polyclonal specifically detects MCAF1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| MCAF1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| activating transcription factor 7 interacting protein, activating transcription factor 7-interacting protein 1, ATF7-interacting protein, ATFa-associated modulator, ATF-interacting protein, ATF-IP, FLJ10139, FLJ10688, hAM, MBD1-containing chromatin associated factor, MCAF1AM, MCAFMBD1-containing chromatin-associated factor 1, MMCP 6, MMCP6, MMCP-6, P621 | |
| Produced in rabbits immunized with E. coli-derived Human MCAF1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 55729 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction