missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mca-KPLGL-Dpa-AR-NH2 Fluorogenic Peptide Substrate
Description
- The peptide substrate contains a highly fluorescent 7-methoxycoumarin group that is efficiently quenched by resonance energy transfer to the 2,4-dinitrophenyl group
- Can be used to measure the activities of peptidases capable of cleaving an amide bond between the fluorescent group and the quencher group, causing an increase in fluorescence
- Purity >95% by HPLC
- The cleavage site is the peptide bond between Gly and Leu
Specifications
Specifications
| Product Line | R&D Systems |
| Quantity | 1 mg |
| Product Type | Mca-Lys-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH2 |
| Storage Requirements | Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?