missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MC5R Recombinant Rabbit Monoclonal Antibody (JE61-09)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544865
This item is not returnable.
View return policy
Description
MC5R Recombinant Monoclonal Antibody for Western Blot, IHC (P)
MC5R, a Melanocortin Receptor, mediates the effects of melanocyte-stimulating hormone (MSH) and adrenocorticotropic hormone (ACTH), and activates adenylate cyclase. This receptor controls multiple functions in the brain and in peripheral tissues, such as thermoregulation, immunomodulation, appetite control, and sexual behavior. Melanocortin 5 receptor has been reported to be expressed in a variety of tissues, especially in pituitary, exocrine glands, reproductive organs, lung, kidney, and esophagus. No human ESTs have been isolated.
Specifications
| MC5R | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P33032, P41149 | |
| MC5R | |
| Synthetic peptide within human MC-2 aa 1-50/325. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JE61-09 | |
| Unconjugated | |
| MC5R | |
| H MC2; MC2; MC-2; MC5; MC5 receptor; MC5R; MC5-R; melanocortin 5 receptor; melanocortin receptor 5; melanocortin-5 receptor | |
| Rabbit | |
| Protein A | |
| RUO | |
| 17203, 4161 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction