missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MC5R Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521906
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa, HepG2. Predicted reactivity: Rhesus Monkey (100%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MC5R, a Melanocortin Receptor, mediates the effects of melanocyte-stimulating hormone (MSH) and adrenocorticotropic hormone (ACTH), and activates adenylate cyclase. This receptor controls multiple functions in the brain and in peripheral tissues, such as thermoregulation, immunomodulation, appetite control, and sexual behavior. Melanocortin 5 receptor has been reported to be expressed in a variety of tissues, especially in pituitary, exocrine glands, reproductive organs, lung, kidney, and esophagus. No human ESTs have been isolated.
Specifications
| MC5R | |
| Polyclonal | |
| Unconjugated | |
| MC5R | |
| H MC2; MC2; MC-2; MC5; MC5 receptor; MC5R; MC5-R; melanocortin 5 receptor; melanocortin receptor 5; melanocortin-5 receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4161 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.18 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P33032 | |
| MC5R | |
| Synthetic peptide corresponding to a region within amino acids 1 and 45 of MC5 Receptor (Uniprot ID#P33032) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction