missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ MC500 Colorimeter Replacement Reagents: Chlorine
For use with MC500 multi-parameter colorimeter
£41.00 - £55.45
Specifications
For Use With (Application) | Water quality testing |
---|---|
For Use With (Equipment) | MC500 multi-parameter colorimeter |
Test Method | DPD |
Type | Chlorine Reagent Set |
Product Code | Brand | Range | Includes | No. of Tests | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Includes | No. of Tests | Price | Quantity & Availability | |||
15351975
|
Lovibond™
517711BT |
0.01 to 6mg/L | DPD No. 1 tablets (100 pack), DPD No. 3 tablets (100 pack) | 100 |
£41.00
Pack of 100 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||
15362015
|
Lovibond™
471056 |
0.02 to 4mg/L | 15mL DPD No. 1 buffer solution, 15mL DPD No. 1 reagent solution, 15mL DPD No. 3 reagent solution | 50 |
£55.45
Pack of 6 |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
For use with MC500 multi-parameter colorimeter
- Parameter: Chlorine
- Method: DPD
Specifications
Water quality testing | |
MC500 multi-parameter colorimeter | |
DPD | |
Chlorine Reagent Set |