Learn More
Invitrogen™ MC3R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595420
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: mouse kidney tissue, COLO320 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene encodes a G-protein-coupled receptor for melanocyte-stimulating hormone and adrenocorticotropic hormone that is expressed in tissues other than the adrenal cortex and melanocytes. This gene maps to the same region as the locus for benign neonatal epilepsy. Mice deficient for this gene have increased fat mass despite decreased food intake, suggesting a role for this gene product in the regulation of energy homeostasis. Mutations in this gene are associated with a susceptibility to obesity in humans.
Specifications
| MC3R | |
| Polyclonal | |
| Unconjugated | |
| MC3R | |
| BMIQ9; MC3; MC3 receptor; MC3R; MC3-R; melanocortin 3 receptor; melanocortin receptor 3; OB20; obesity quantitative trait locus; OQTL | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17201, 4159 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P33033, P41968 | |
| MC3R | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human MC3 Receptor (91-121aa NALETIMIAIVHSDYLTFEDQFIQHMDNIFD). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.