missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MC1R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104251
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MSHR.
MC1R, a Melanocortin Receptor, mediates the effects of melanocyte-stimulating hormone (MSH) and adrenocorticotropic hormone (ACTH). MSH and its receptor, MC1R, are key regulators of melanogenesis. They control the relative proportions of red pheomelanin and black photoprotective eumelanin in the skin by increasing the production of eumelanin. Non-functional, truncated forms of the receptor lead to lighter coat color in animals. In contrast, constitutively active receptors lead to darker color.
Specifications
| MC1R | |
| Polyclonal | |
| Unconjugated | |
| Mc1r | |
| alpha melanocyte stimulating hormone receptor; CMM5; e; extension recessive yellow; MC1 receptor; MC1R; MC1-R; melanocortin 1 receptor; melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor); melanocortin receptor 1; melanocortin-1 receptor; melanocyte-stimulating hormone receptor; melanotropin receptor; MSHR; MSH-R; Mshra; SHEP2; Tob; tobacco darkening | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4157 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q01726 | |
| Mc1r | |
| A synthesized peptide derived from human MC1R(Accession Q01726), corresponding to amino acid residues N279-W317. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction