missing translation for 'onlineSavingsMsg'
Learn More

Mc-MMAD, MedChemExpress

Product Code. 16456788
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16456788 10 mg 10mg
16429337 5 mg 5mg
16421549 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16456788 Supplier Medchem Express Supplier No. HY15740
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Mc-MMAD is a protective group (maleimidocaproyl)-conjugated MMAD. MMAD is a potent tubulin inhibitor. Mc-MMAD is a drug-linker conjugate for ADC.

Mc-MMAD is a protective group (maleimidocaproyl)-conjugated MMAD. MMAD is a potent tubulin inhibitor. Mc-MMAD is a drug-linker conjugate for ADC.

Specifications

Chemical Name or Material Mc-MMAD
CAS 1401963-15-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C51H77N7O9S
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (103.71 mM)
SMILES O=C1N(CCCCCC(N(C)[C@H](C(N[C@@H](C(C)C)C(N(C)[C@]([C@@H](C)CC)([H])[C@@H](CC(N2[C@H]([C@H](OC)[C@H](C(N[C@@H](CC3=CC=CC=C3)C4=NC=CS4)=O)C)CCC2)=O)OC)=O)=O)C(C)C)=O)C(C=C1)=O
Molecular Weight (g/mol) 964.26
Formula Weight 964.26
Percent Purity 98.5%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.