missing translation for 'onlineSavingsMsg'
Learn More

MBTPS1 Antibody (2E6), Novus Biologicals™

Product Code. 18392999 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392999 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392999 Supplier Novus Biologicals Supplier No. H00008720M07

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MBTPS1 Monoclonal antibody specifically detects MBTPS1 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBTPS1
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2E6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Alias EC 3.4.21.112, Endopeptidase S1P, KIAA0091MGC138712, membrane-bound transcription factor peptidase, site 1, membrane-bound transcription factor protease, site 1, PCSK8, S1PMGC138711, site-1 protease, SKI1, SKI-1membrane-bound transcription factor site-1 protease, subtilisin/kexin isozyme-1, Subtilisin/kexin-isozyme 1
Host Species Mouse
Immunogen MBTPS1 (NP_957720.1, 246 a.a. ~ 355 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GLGHGTFVAGVIASMRECQGFAPDAELHIFRVFTNNQVSYTSWFLDAFNYAILKKIDVLNLSIGGPDFMDHPFVDKVWELTANNVIMVSAIGNDGPLYGTLNNPADQMDV
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8720
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.