missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBQ-167, MedChemExpress
Description
MBQ-167 is a dual Rac/Cdc42 inhibitor, with IC50s of 103 nM for Rac 1/2/3 and 78 nM for Cdc42 in MDA-MB-231 cells, respectively.
Specifications
Specifications
| Chemical Name or Material | MBQ-167 |
| CAS | 2097938-73-1 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C22H18N4 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 155 mg/mL (458.02 mM; Need ultrasonic) |
| SMILES | CCN1C2=C(C3=C1C=CC=C3)C=C(N4N=NC=C4C5=CC=CC=C5)C=C2 |
| Molecular Weight (g/mol) | 338.41 |
| Formula Weight | 338.41 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?