Learn More
Invitrogen™ MBP Monoclonal Antibody (MBP101)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA110837
Product Code. 11563443
  • £732.00 / 500µg
Description
MA1-10837 detects Myelin Basic Protein from human, non-human primate, rabbit, sheep, goat, rat, and mouse samples. MA1-10837 has been successfully used in ELISA, Western blot, and immunohistochemistry applications. The MA1-10837 immunogen is purified human myelin basic protein.
The protein encoded by the classic MBP gene is a major constituent of the myelin sheath of oligodendrocytes and Schwann cells in the nervous system. However, MBP-related transcripts are also present in the bone marrow and the immune system. These mRNAs arise from the long MBP gene (otherwise called 'Golli-MBP') that contains 3 additional exons located upstream of the classic MBP exons. Alternative splicing from the Golli and the MBP transcription start sites gives rise to 2 sets of MBP-related transcripts and gene products. The Golli mRNAs contain 3 exons unique to Golli-MBP, spliced in-frame to 1 or more MBP exons. They encode hybrid proteins that have N-terminal Golli aa sequence linked to MBP aa sequence. The second family of transcripts contain only MBP exons and produce the well characterized myelin basic proteins. This complex gene structure is conserved among species suggesting that the MBP transcription unit is an integral part of the Golli transcription unit and that this arrangement is important for the function and/or regulation of these genes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MBP
Monoclonal
1.7 mg/mL
PBS with no preservative; pH 7.4
P02686, P02688, P04370, P25274
MBP
Purified human myelin basic protein.
500 μg
Primary
Goat, Human, Mouse, Monkey, Sheep, Rabbit, Rat
Antibody
IgG2b
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
MBP101
Unconjugated
MBP
20 kDa microtubule-stabilizing protein; C76307; golli mbp; Golli-mbp; Golli-MBP; myelin basic protein; Golli-Mbp; myelin basic protein; myelin basic protein S; Hmbpr; MBP; MBP S; Mbps; MGC99675; microtubule-stabilizing protein; mld; MOBP; Myelin A1 protein; myelin basic protein; myelin basic protein Golli-mbp; myelin basic protein S; myelin deficient; myelin membrane encephalitogenic protein; Myelin P1 protein; myelin-associated oligodendrocyte basic protein; R75289; shi; shiverer; Unknown (protein for IMAGE:7984318); unnamed protein product
Mouse
Protein G
RUO
100344447, 101109407, 106503529, 17196, 24547, 4155
-20°C, Avoid Freeze/Thaw Cycles