missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Antibody (rMBP/9301), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23738-100ug
This item is not returnable.
View return policy
Description
MBP Monoclonal antibody specifically detects MBP in Human, Rat, Porcine, Chicken, Guinea Pig, Mammalia, Rabbit, Sheep samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MBP | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
| Recombinant fragment (around aa1-157) of human MBP protein (exact sequence is proprietary) | |
| 100 μg | |
| Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers | |
| 4155 | |
| Store at 4°C. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| rMBP/9301 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat, Pig, Chicken, Guinea Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction