missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MBP Polyclonal antibody specifically detects MBP in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | MBP |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein |
| Host Species | Rabbit |
| Immunogen | Reacts with residues 49-62 [DRGAPKRGSGKDSH] of the 33 kDa human, mouse and rat MBP protein. Antibody detects primarily the 22 kDa isoform. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?