Learn More
MBP Antibody (CL2829) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46632PE
Product Code. 30293924
  • £567.00 / 0.10mL
Description
MBP Monoclonal antibody specifically detects MBP in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MBP
Monoclonal
PE
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein
This antibody was developed using a recombinant protein derived from P02686, with the exact immunogen sequence remaining proprietary.
0.1 mL
Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers
4155
Store at 4⪚C in the dark.
IgG1
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL2829
PBS
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified