missing translation for 'onlineSavingsMsg'
Learn More

MBP Antibody (CL2819), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-46630-25ul

 View more versions of this product

Product Code. 18609285

  • £272.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MBP Monoclonal antibody specifically detects MBP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MBP
Monoclonal
Unconjugated
PBS (pH 7.2), 40% Glycerol
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence DENPVVHFFKNIVTPRTPPPSQGKGRGLSLSRFSWGAEGQRPGFGYGGRASDYKSAHKGFKGVDAQGTLSKIFKLGGRDSRSGSPM
25 μL
Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers
4155
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
CL2819
Western Blot 1 μg/mL, Immunohistochemistry 1:5000 - 1:10000, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:5000 - 1:10000
P02686
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.