missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MBOAT7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563224
This item is not returnable.
View return policy
Description
Immunogen sequence: LGGGSPSRRK AASQPTSLAP EKLREE Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.
MBOAT7 belongs to the membrane-bound acyltransferase family. It is involved in arachidonate recycling, thus regulating free arachidonic acid levels and leukotriene synthesis in neutrophils.
Specifications
| MBOAT7 | |
| Polyclonal | |
| Unconjugated | |
| MBOAT7 | |
| 1-acylglycerophosphatidylinositol O-acyltransferase; 5730589L02Rik; BB1; Bladder and breast carcinoma-overexpressed gene 1 protein; h-mboa-7; hMBOA-7; Leng4; leukocyte receptor cluster (LRC) member 4; leukocyte receptor cluster member 4; LPIAT; Lpiat1; LPLAT 7; LRC4; lysophosphatidylinositol acyltransferase; Lysophosphatidylinositol acyltransferase 1; Lysophospholipid acyltransferase 7; lyso-PI acyltransferase; malignant cell expression-enhanced gene/tumor progression-enhanced; mBB1; MBOA7; Mboat7; membrane bound O-acyltransferase domain containing 7; membrane-bound O-acyltransferase domain-containing protein 7; m-mboa-7; OACT7; O-acyltransferase domain-containing protein 7; RGD1306945 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79143 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96N66 | |
| MBOAT7 | |
| Recombinant protein corresponding to Human MBOAT7. Recombinant protein control fragment (Product #RP-101661). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction