missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MBOAT4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532916
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan (100%) Gibbon, Monkey, Galago, Panda, Rabbit (93%) Mouse, Bat, Horse (86%).
Mediates the octanoylation of ghrelin at 'Ser-3'. Can use a variety of fatty acids as substrates including octanoic acid, decanoic acid and tetradecanoic acid.
Specifications
| MBOAT4 | |
| Polyclonal | |
| Unconjugated | |
| MBOAT4 | |
| EMBL:ACB05875.1}; FKSG89; Ghrelin O-acyltransferase; ghrelin O-acyltransferase {ECO:0000303; Gm171; GOAT; Mboat4; mboat4 {ECO:0000312; membrane bound O-acyltransferase domain containing 4; membrane-bound O-acyltransferase domain-containing protein 4; membrane-bound O-acyltransferase domain-containing protein 4 {ECO:0000312; OACT4; O-acyltransferase (membrane bound) domain containing 4; O-acyltransferase domain-containing protein 4; PubMed:18443287}; RGD:1566335}; RGD1566335 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 619373 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q96T53 | |
| MBOAT4 | |
| Synthetic 14 amino acid peptide from internal region of human MBOAT4. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction