missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MBOAT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553238
This item is not returnable.
View return policy
Description
Immunogen sequence: TQRRKNTHEN IQLSQSKKFD EGENSLGQNS FSTTNNVCNQ NQEIASRHSS LKQ Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 70%.
Acyltransferase which mediates the conversion of lysophosphatidylethanolamine (1-acyl-sn-glycero-3- phosphoethanolamine or LPE) into phosphatidylethanolamine (1,2- diacyl-sn-glycero-3-phosphoethanolamine or PE) (LPEAT activity). Catalyzes also the acylation of lysophosphatidic acid (LPA) into phosphatidic acid (PA) (LPAAT activity). Has also a very weak lysophosphatidylcholine acyltransferase (LPCAT activity). Prefers oleoyl-CoA as the acyl donor. Lysophospholipid acyltransferases (LPLATs) catalyze the reacylation step of the phospholipid remodeling pathway also known as the Lands cycle.
Specifications
| MBOAT2 | |
| Polyclonal | |
| Unconjugated | |
| MBOAT2 | |
| 1-acylglycerophosphate O-acyltransferase; 1-acylglycerophosphocholine O-acyltransferase; 1-acylglycerophosphoethanolamine O-acyltransferase; 231n7; 2810049G06Rik; AU022889; AW547221; LPAAT; LPC acyltransferase 4; LPCAT; Lpcat4; LPEAT; LPLAT 2; lyso-PA acyltransferase; lyso-PC acyltransferase 4; lyso-PE acyltransferase; lysophosphatidic acid acyltransferase; Lysophosphatidylcholine acyltransferase; Lysophosphatidylcholine acyltransferase 4; lysophosphatidylethanolamine acyltransferase; Lysophospholipid acyltransferase 2; MBOAT2; membrane bound O-acyltransferase domain containing 2; membrane-bound O-acyltransferase domain-containing protein 2; mLPCAT4; Moact2; OACT2; O-acyltransferase (membrane bound) domain containing 2; O-acyltransferase domain-containing protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 129642 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZWT7 | |
| MBOAT2 | |
| Recombinant protein corresponding to Human MBOAT2. Recombinant protein control fragment (Product #RP-91250). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction