missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MBNL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551603
This item is not returnable.
View return policy
Description
Immunogen sequence: AQMSSLGSFP MTPSIPANPP MAFNPYIPHP GMGLVPAELV PNTPVLIPGN PPLAMPGAVG PKLMRSDKLE VCREFQRGNC TRGENDCRYA HPTDASMIEA SDNTVTICMD YIKGRCSREK CKYFHPPAHL QARLKAAHHQ MNHSAAS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.
This gene encodes a member of the muscleblind-like family of proteins. The encoded protein may function in regulation of alternative splicing and may play a role in the pathophysiology of myotonic dystrophy. Alternatively spliced transcript variants have been described.
Specifications
| MBNL3 | |
| Polyclonal | |
| Unconjugated | |
| Mbnl3 | |
| A530038J18Rik; AI661274; CHCR; Cys3His CCG1-required protein; E430034C16Rik; MBLX; MBLX39; Mbnl3; MBXL; muscleblind like splicing regulator 3; muscleblind-like 3; muscleblind-like 3 (Drosophila); muscleblind-like protein 3; muscleblind-like splicing regulator 3; muscleblind-like X-linked protein; Protein HCHCR; Protein MCHCR; RGD1565834; RP5-842K24.3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55796 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NUK0 | |
| Mbnl3 | |
| Recombinant protein corresponding to Human MBNL3. Recombinant protein control fragment (Product #RP-92116). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction