missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ MBM™-4 Melanocyte Growth Basal Medium-4
Click to view available options
Quantity:
500 mL
Unit Size:
Each
Description
- Basal medium for supporting melanocyte growth.
- Can be used with CC-4465 (melanocyte growth medium SingleQuots™ Supplements and Growth Factors).
- The full MGM™ - 4 BulletKit™ consisting of basal medium and supplements can be purchased using the catalog number CC-3249.
- Provides flexibility to add or omit components as research needs dictate.
- ET-3 Growth Supplement, an additional growth factor required for adult melanocyte growth, is sold separately to supplement the MGM™-4 BulletKit™ for a complete formulation specifically designed for growing adult melanocytes.
- ET-3 offers enhanced growth and plating advantages in addition to limiting other cell type contamination in culture.
Specifications
Specifications
| Cell Type | Melanocytes |
| Product Type | Basal Medium |
| Quantity | 500 mL |
| pH | 7.25 to 7.65 |
| Color | Red |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction